This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Os03623096_m1
See other LOC4329635 GE Assays ›
Gene Symbol:
LOC4329635
Gene Name:
zinc finger transcription factor YY1
Gene Aliases:
OJ1004_H01.19, OSNPB_020551900
Chromosome Location:
-
Species:
Rice
Species Specific ID (Flybase ID):
-
Assay ID Os03623096_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC4329635
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015772203.2 XP_015627689.1 2 - 3 405 59
XM_015772204.2 XP_015627690.1 2 - 3 405 59
XM_015772205.2 XP_015627691.1 2 - 3 359 59
GenBank mRNA AK103760.1 - 2 - 3 411 59

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Oryza sativa Probe spans exons

Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-97tfg:80/100.66.76.242:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline