This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh00359559_m1
See other SYNCRIP GE Assays ›
Gene Symbol:
SYNCRIP
Gene Name:
synaptotagmin binding cytoplasmic RNA interacting protein
Gene Aliases:
-
Chromosome Location:
Chr.4: 87592497 - 87628766 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh00359559_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SYNCRIP
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001193845.2 NP_001180774.1 1823 75
XM_001089294.4 XP_001089294.1 1405 75
XM_015136873.2 XP_014992359.1 1449 75
XM_015136874.2 XP_014992360.1 1823 75
XM_015136876.2 XP_014992362.1 1449 75
XM_015136877.2 XP_014992363.1 1271 75
XM_015136878.2 XP_014992364.1 1271 75
XM_015136879.2 XP_014992365.1 1199 75
XM_028846654.1 XP_028702487.1 1449 75
GenBank mRNA JU319741.1 - 1316 75
JU319742.1 - 1476 75
JU471299.1 - 1796 75
JU473997.1 - 1796 75
JV044032.1 - 1819 75
JV045925.1 - 1819 75
JV600201.1 - 1626 75
JV640271.1 - 1788 75
JV703629.1 - 1316 75

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Macaca mulatta

Panther Classification:

Molecular Function -

RNA metabolism protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0