Assay Details
| Gene Symbol: RYBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028844364.1 | XP_028700197.1 | 373 | 83 | ||
| GenBank mRNA | JU327892.1 | - | 150 | 83 | ||
| JU474935.1 | - | 245 | 83 | |||
| JV045003.1 | - | 258 | 83 | |||
| JV602413.1 | - | 268 | 83 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

RYBP
Genomic Map