This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh00414732_m1
See other NAA38 GE Assays ›
Gene Symbol:
NAA38 CYB5D1
Gene Name:
N-alpha-acetyltransferase 38, NatC auxiliary subunit
cytochrome b5 domain containing 1
Gene Aliases:
LSMD1
EGK_08149
Chromosome Location:
Chr.16: 7649156 - 7650363 on Build Mmul_10
Chromosome Location:
Chr.16: 7650203 - 7706546 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
-
Assay ID Rh00414732_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NAA38
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_028835982.1 XP_028691815.1 692 105
GenBank mRNA JU328525.1 - 144 105
JV073339.1 - 353 105
Gene Symbol: CYB5D1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA JV663393.1 - 1424 105

Back To Top

More Information


Additional Information:

This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Macaca mulatta

Panther Classification:

Molecular Function -

RNA splicing factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rr9bx:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0