This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh00697699_m1
See other ETFBKMT GE Assays ›
Gene Symbol:
ETFBKMT
Gene Name:
electron transfer flavoprotein subunit beta lysine methyltransferase
Gene Aliases:
C11H12orf72, C12orf72
Chromosome Location:
Chr.11: 32774821 - 32860273 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh00697699_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ETFBKMT
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001193718.2 NP_001180647.1 658 146
XM_015151358.2 XP_015006844.1 748 146
XM_015151359.2 XP_015006845.1 692 146
XM_028828805.1 XP_028684638.1 973 146
XM_028828806.1 XP_028684639.1 749 146
XM_028828807.1 XP_028684640.1 749 146

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Macaca mulatta

Panther Classification:

Molecular Function -

methyltransferase transferase metabolite interconversion enzyme

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0