This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh00855957_m1
See other PRSS37 GE Assays ›
Gene Symbol:
PRSS37
Gene Name:
serine protease 37
Gene Aliases:
EGK_14216
Chromosome Location:
Chr.3: 168223715 - 168251981 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh00855957_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PRSS37
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001194861.3 NP_001181790.2 545 88
XM_001083561.3 XP_001083561.2 260 88
XM_015135073.2 XP_014990559.1 260 88
XM_028844798.1 XP_028700631.1 307 88

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Recommended Assay Macaca mulatta

Panther Classification:

Molecular Function -

protease serine protease protein modifying enzyme

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-9hc65:80/100.66.78.230:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0