This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh00895275_m1
See other PCTP GE Assays ›
Gene Symbol:
PCTP
Gene Name:
phosphatidylcholine transfer protein
Gene Aliases:
-
Chromosome Location:
Chr.16: 40934817 - 41043043 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh00895275_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PCTP
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_001104194.4 XP_001104194.2 610 97
XM_028836326.1 XP_028692159.1 711 97
XM_028836327.1 XP_028692160.1 711 97
XM_028836328.1 XP_028692161.1 739 97
XM_028836329.1 XP_028692162.1 711 97
XM_028836330.1 XP_028692163.1 711 97
XM_028836331.1 XP_028692164.1 711 97
XM_028836332.1 XP_028692165.1 711 97
XM_028836333.1 XP_028692166.1 610 97
XM_028836334.1 XP_028692167.1 610 97

Back To Top

More Information


Additional Information:

This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.

Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Macaca mulatta

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline