This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh00895475_g1
See other AKR1A1 GE Assays ›
Gene Symbol:
AKR1A1
Gene Name:
aldo-keto reductase family 1 member A1
Gene Aliases:
EGK_00701
Chromosome Location:
Chr.1: 178852849 - 178872515 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh00895475_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AKR1A1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001261358.1 NP_001248287.1 820 106
XM_015138324.2 XP_014993810.2 949 106
XM_015138328.2 XP_014993814.2 931 106
XM_015138335.2 XP_014993821.2 834 106
XM_028852936.1 XP_028708769.1 950 106
GenBank mRNA JU322102.1 - 1091 106
JU471778.1 - 1097 106
JV044459.1 - 820 106
JV566770.1 - 852 106
JV602285.1 - 858 106

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta

Panther Classification:

Molecular Function -

oxidoreductase reductase metabolite interconversion enzyme

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0