This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh00909803_m1
See other NAPG GE Assays ›
Gene Symbol:
NAPG
Gene Name:
NSF attachment protein gamma
Gene Aliases:
-
Chromosome Location:
Chr.18: 71455740 - 71488187 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh00909803_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NAPG
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001258101.1 NP_001245030.1 548 113
XM_028837743.1 XP_028693576.1 681 113
GenBank mRNA JU336918.1 - 556 113
JU477652.1 - 548 113
JU477653.1 - 548 113
JV632198.1 - 583 113

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Recommended Assay Macaca mulatta

Panther Classification:

Molecular Function -

membrane traffic protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-f9vrx:80/100.66.79.34:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0