This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh00918623_m1
See other MOV10 GE Assays ›
Gene Symbol:
MOV10
Gene Name:
Mov10 RISC complex RNA helicase
Gene Aliases:
-
Chromosome Location:
Chr.1: 111512063 - 111538655 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh00918623_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MOV10
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001261223.1 NP_001248152.1 3033 72
XM_015147103.2 XP_015002589.1 3214 72
XM_015147118.2 XP_015002604.1 3963 72
XM_028843048.1 XP_028698881.1 3137 72
XM_028843052.1 XP_028698885.1 3435 72
XM_028843059.1 XP_028698892.1 3314 72
XM_028843061.1 XP_028698894.1 3413 72
GenBank mRNA JU472481.1 - 2745 72
JV043925.1 - 3049 72
JV043926.1 - 3033 72
JV636164.1 - 3054 72
JV636798.1 - 3070 72

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Macaca mulatta

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0