This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh00936498_m1
See other MDH1 GE Assays ›
Gene Symbol:
MDH1
Gene Name:
malate dehydrogenase 1
Gene Aliases:
-
Chromosome Location:
Chr.13: 45173984 - 45192344 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh00936498_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MDH1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001320104.1 NP_001307033.1 870 78
NM_001320105.1 NP_001307034.1 870 78
NM_001320106.1 NP_001307035.1 1031 78
GenBank mRNA CN647359.1 - 524 78
JU327268.1 - 539 78
JU475632.1 - 853 78
JV044980.1 - 853 78
JV490880.1 - 850 78
JV545249.1 - 830 78
JV635880.1 - 853 78

Back To Top

More Information


Additional Information:

This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.

Set Membership:

Amplicon length between 71 and 85 Recommended Assay Macaca mulatta

Panther Classification:

Molecular Function -

dehydrogenase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline