This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh00938057_m1
See other SLC7A6 GE Assays ›
Gene Symbol:
SLC7A6
Gene Name:
solute carrier family 7 member 6
Gene Aliases:
EGK_12923
Chromosome Location:
Chr.20: 55490447 - 55532877 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh00938057_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SLC7A6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015126466.2 XP_014981952.1 1576 84
XM_015126467.2 XP_014981953.1 2194 84
XM_015126468.2 XP_014981954.1 1345 84
XM_028841550.1 XP_028697383.1 1597 84
XM_028841551.1 XP_028697384.1 1720 84
GenBank mRNA JU337005.1 - 1329 84
JU337006.1 - 1349 84
JU471058.1 - 1351 84
JV048633.1 - 1349 84

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Macaca mulatta

Panther Classification:

Molecular Function -

transporter

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline