This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh00956611_m1
See other ELAVL4 GE Assays ›
Gene Symbol:
ELAVL4
Gene Name:
ELAV like RNA binding protein 4
Gene Aliases:
EGK_00744
Chromosome Location:
Chr.1: 174265739 - 174420440 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh00956611_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ELAVL4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001258180.1 NP_001245109.1 516 91
XM_015139107.2 XP_014994593.1 499 91
XM_015139118.2 XP_014994604.1 493 91
XM_015139127.2 XP_014994613.1 691 91
XM_015139130.2 XP_014994616.1 511 91
XM_015139139.2 XP_014994625.1 620 91
XM_015139148.2 XP_014994634.1 488 91
XM_015139161.2 XP_014994647.1 693 91
XM_015139169.2 XP_014994655.1 510 91
XM_028844296.1 XP_028700129.1 410 91
XM_028844310.1 XP_028700143.1 707 91
XM_028844334.1 XP_028700167.1 707 91
GenBank mRNA JU333292.1 - 516 91
JU333293.1 - 516 91
JV459479.1 - 520 91
JV608269.1 - 523 91

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Recommended Assay Macaca mulatta

Panther Classification:

Molecular Function -

translation initiation factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0