This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh00960672_mH
See other UBE2D3 GE Assays ›
Gene Symbol:
UBE2D3
Gene Name:
ubiquitin conjugating enzyme E2 D3
Gene Aliases:
-
Chromosome Location:
Chr.5: 101194610 - 101267834 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh00960672_mH
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: UBE2D3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001261204.1 NP_001248133.1 555 76
XM_015138841.2 XP_014994327.1 571 76
XM_015138842.2 XP_014994328.1 618 76
XM_015138845.2 XP_014994331.1 664 76
XM_015138847.2 XP_014994333.1 795 76
XM_015138848.2 XP_014994334.1 306 76
XM_015138849.2 XP_014994335.1 459 76
XM_028848100.1 XP_028703933.1 744 76
GenBank mRNA JU328417.1 - 128 76
JU471195.1 - 778 76
JU477051.1 - 597 76
JU477052.1 - 980 76
JV048140.1 - 555 76
JV636355.1 - 564 76
JV660447.1 - 572 76

Back To Top

More Information


Additional Information:

This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.

Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Macaca mulatta

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline