This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh00964146_m1
See other HINFP GE Assays ›
Gene Symbol:
HINFP
Gene Name:
histone H4 transcription factor
Gene Aliases:
EGK_06980
Chromosome Location:
Chr.14: 112185814 - 112204471 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh00964146_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HINFP
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001194253.2 NP_001181182.1 1232 77
XM_001103929.4 XP_001103929.1 1259 77
XM_015115896.2 XP_014971382.1 1265 77
XM_028832608.1 XP_028688441.1 1330 77
XM_028832609.1 XP_028688442.1 1257 77
XM_028832612.1 XP_028688445.1 1310 77
XM_028832613.1 XP_028688446.1 1312 77
GenBank mRNA JU321085.1 - 1165 77
JU475078.1 - 1173 77
JV043452.1 - 1181 77
JV463997.1 - 1185 77

Back To Top

More Information


Additional Information:

This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.

Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Macaca mulatta

Panther Classification:

Molecular Function -

C2H2 zinc finger transcription factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline