This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh00976243_m1
See other ESRRG GE Assays ›
Gene Symbol:
ESRRG
Gene Name:
estrogen related receptor gamma
Gene Aliases:
EGK_01652
Chromosome Location:
Chr.1: 73998664 - 74592891 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh00976243_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ESRRG
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_028853225.1 XP_028709058.1 2002 76
XM_028853228.1 XP_028709061.1 1956 76
XM_028853232.1 XP_028709065.1 1390 76
XM_028853236.1 XP_028709069.1 1630 76
XM_028853238.1 XP_028709071.1 1147 76
XM_028853242.1 XP_028709075.1 1271 76
XM_028853248.1 XP_028709081.1 1305 76
XM_028853254.1 XP_028709087.1 1605 76
XM_028853260.1 XP_028709093.1 1193 76
XM_028853266.1 XP_028709099.1 1363 76
XM_028853270.1 XP_028709103.1 1244 76
XM_028853273.1 XP_028709106.1 1654 76
XM_028853279.1 XP_028709112.1 1932 76
XM_028853286.1 XP_028709119.1 1865 76

Back To Top

More Information


Additional Information:

This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.

Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Macaca mulatta

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-55ff9ddd88-xhqb4:80/100.66.79.154:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0