This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh00978041_m1
See other HDAC11 GE Assays ›
Gene Symbol:
HDAC11
Gene Name:
histone deacetylase 11
Gene Aliases:
-
Chromosome Location:
Chr.2: 144875316 - 144903615 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh00978041_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HDAC11
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001265655.1 NP_001252584.1 568 72
XM_015130111.2 XP_014985597.1 982 72
XM_015130112.2 XP_014985598.1 531 72
XM_015130113.2 XP_014985599.1 504 72
XM_015130114.2 XP_014985600.1 1003 72
XM_028843060.1 XP_028698893.1 579 72
XM_028843062.1 XP_028698895.1 528 72
XM_028843063.1 XP_028698896.1 504 72
XM_028843064.1 XP_028698897.1 859 72
XM_028843065.1 XP_028698898.1 462 72
XM_028843066.1 XP_028698899.1 838 72
XM_028843067.1 XP_028698900.1 605 72
GenBank mRNA JU327416.1 - 531 72
JV636381.1 - 568 72
JV696806.1 - 614 72

Back To Top

More Information


Additional Information:

This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.

Set Membership:

Amplicon length between 71 and 85 Recommended Assay Macaca mulatta

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline