This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh00992348_m1
See other KMT5B GE Assays ›
Gene Symbol:
KMT5B
Gene Name:
lysine methyltransferase 5B
Gene Aliases:
EGK_05912, SUV420H1
Chromosome Location:
Chr.14: 6556415 - 6616082 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh00992348_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: KMT5B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001260793.1 NP_001247722.1 884 88
XM_028832661.1 XP_028688494.1 1157 88
XM_028832662.1 XP_028688495.1 1088 88
XM_028832663.1 XP_028688496.1 866 88
XM_028832664.1 XP_028688497.1 1075 88
XM_028832666.1 XP_028688499.1 710 88
XM_028832667.1 XP_028688500.1 446 88
XM_028832668.1 XP_028688501.1 1157 88
GenBank mRNA JU334792.1 - 777 88
JU471551.1 - 913 88
JV044469.1 - 884 88
JV453008.1 - 738 88

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Recommended Assay Macaca mulatta

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0