This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh00999645_m1
See other ARHGAP19 GE Assays ›
Gene Symbol:
ARHGAP19
Gene Name:
Rho GTPase activating protein 19
Gene Aliases:
-
Chromosome Location:
Chr.9: 97661900 - 97740399 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh00999645_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ARHGAP19
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001266583.1 NP_001253512.1 1495 73
XM_015147777.2 XP_015003263.2 1541 73
XM_015147778.2 XP_015003264.2 1495 73
XM_015147779.2 XP_015003265.2 1427 73
XM_015147781.2 XP_015003267.2 1294 73
XM_015147783.2 XP_015003269.2 1321 73
XM_028853395.1 XP_028709228.1 1322 73
XM_028853396.1 XP_028709229.1 1236 73
GenBank mRNA JU336810.1 - 1496 73
JU477561.1 - 1495 73
JU477562.1 - 1495 73
JV048498.1 - 1495 73

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Macaca mulatta

Panther Classification:

Molecular Function -

GTPase-activating protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0