This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh01011060_m1
See other BATF3 GE Assays ›
Gene Symbol:
BATF3
Gene Name:
basic leucine zipper ATF-like transcription factor 3
Gene Aliases:
-
Chromosome Location:
Chr.1: 70170982 - 70186692 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh01011060_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: BATF3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_001107737.4 XP_001107737.1 445 78
XM_015117211.2 XP_014972697.1 409 78
XM_028853023.1 XP_028708856.1 446 78
GenBank mRNA JU489332.1 - 108 78
JV655478.1 - 133 78

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Macaca mulatta

Panther Classification:

Molecular Function -

basic leucine zipper transcription factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0