This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh01017682_m1
See other MBD4 GE Assays ›
Gene Symbol:
MBD4
Gene Name:
methyl-CpG binding domain 4, DNA glycosylase
Gene Aliases:
-
Chromosome Location:
Chr.2: 147055810 - 147068052 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh01017682_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MBD4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015130159.2 XP_014985645.2 701 76
XM_015130160.2 XP_014985646.2 540 76
XM_015130161.2 XP_014985647.2 540 76
XM_015130163.2 XP_014985649.2 540 76
XM_028844501.1 XP_028700334.1 540 76
GenBank mRNA JU336958.1 - 471 76
JU336959.1 - 471 76
JU477693.1 - 507 76
JU477694.1 - 507 76
JV048611.1 - 495 76
JV664723.1 - 508 76

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Macaca mulatta

Panther Classification:

Molecular Function -

DNA metabolism protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7gdsv:80/100.66.79.88:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0