This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh01042039_m1
See other ATG4C GE Assays ›
Gene Symbol:
ATG4C
Gene Name:
autophagy related 4C cysteine peptidase
Gene Aliases:
-
Chromosome Location:
Chr.1: 161550223 - 161669480 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh01042039_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ATG4C
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001257887.1 NP_001244816.1 993 81
XM_028828297.1 XP_028684130.1 1153 81
XM_028828330.1 XP_028684163.1 1111 81
XM_028828372.1 XP_028684205.1 1351 81
XM_028828409.1 XP_028684242.1 1097 81
XM_028828436.1 XP_028684269.1 1303 81
XM_028828458.1 XP_028684291.1 1142 81
XM_028828500.1 XP_028684333.1 1907 81
XM_028828523.1 XP_028684356.1 1138 81
XM_028828553.1 XP_028684386.1 1153 81
XM_028828585.1 XP_028684418.1 1151 81
XM_028828619.1 XP_028684452.1 1111 81
XM_028828654.1 XP_028684487.1 1297 81
XM_028828672.1 XP_028684505.1 1152 81
XM_028828711.1 XP_028684544.1 1152 81
GenBank mRNA JU322374.1 - 993 81
JU472960.1 - 1053 81

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Recommended Assay Macaca mulatta

Panther Classification:

Molecular Function -

cysteine protease

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0