This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh01050239_m1
See other EIF5A GE Assays ›
Gene Symbol:
EIF5A
Gene Name:
eukaryotic translation initiation factor 5A
Gene Aliases:
EGK_08114
Chromosome Location:
Chr.16: 7082286 - 7088073 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh01050239_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: EIF5A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015118463.2 XP_014973949.1 538 68
XM_015118465.2 XP_014973951.1 794 68
XM_015118466.2 XP_014973952.1 589 68
XM_015118467.2 XP_014973953.1 561 68
XM_015118470.2 XP_014973956.1 630 68
XM_028835959.1 XP_028691792.1 475 68
GenBank mRNA JV608243.1 - 543 68
JV636008.1 - 500 68

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Macaca mulatta

Panther Classification:

Molecular Function -

translation initiation factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0