This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh01061427_g1
See other RBPJ GE Assays ›
Gene Symbol:
RBPJ
Gene Name:
recombination signal binding protein for immunoglobulin kappa J region
Gene Aliases:
-
Chromosome Location:
Chr.5: 25130889 - 25400522 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh01061427_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RBPJ
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001260693.1 NP_001247622.1 800 100
XM_015138099.2 XP_014993585.1 650 100
XM_015138100.2 XP_014993586.1 724 100
XM_015138101.2 XP_014993587.1 822 100
XM_028847960.1 XP_028703793.1 953 100
GenBank mRNA JU334354.1 - 687 100
JU334355.1 - 599 100
JU475456.1 - 692 100
JV046967.1 - 800 100
JV046968.1 - 617 100

Back To Top

More Information


Additional Information:

This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.

Set Membership:

Amplicon length between 86 and 100 Recommended Assay Macaca mulatta

Panther Classification:

Molecular Function -

DNA-binding transcription factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0