This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh01065382_m1
See other GABPB1 GE Assays ›
Gene Symbol:
GABPB1
Gene Name:
GA binding protein transcription factor subunit beta 1
Gene Aliases:
EGK_17484, GABPAP
Chromosome Location:
Chr.7: 26686531 - 26765929 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh01065382_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GABPB1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001194637.2 NP_001181566.1 699 88
XM_001114320.4 XP_001114320.1 3134 88
XM_015142201.2 XP_014997687.1 635 88
XM_015142202.2 XP_014997688.1 3158 88
XM_015142208.2 XP_014997694.1 3158 88
XM_028850500.1 XP_028706333.1 299 88
XM_028850501.1 XP_028706334.1 300 88
XM_028850502.1 XP_028706335.1 3158 88
XM_028850503.1 XP_028706336.1 299 88
XM_028850504.1 XP_028706337.1 300 88
XM_028850505.1 XP_028706338.1 3158 88
GenBank mRNA JU472364.1 - 676 88
JU472365.1 - 676 88
JV046658.1 - 681 88
JV046659.1 - 681 88

Back To Top

More Information


Additional Information:

This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.

Set Membership:

Amplicon length between 86 and 100 3' Most Recommended Assay Macaca mulatta

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0