This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh01086566_g1
See other GTF2F1 GE Assays ›
Gene Symbol:
GTF2F1
Gene Name:
general transcription factor IIF subunit 1
Gene Aliases:
-
Chromosome Location:
Chr.19: 6313215 - 6327750 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh01086566_g1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GTF2F1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001260830.1 NP_001247759.1 1339 86
GenBank mRNA JU334180.1 - 1301 86
JU334181.1 - 1301 86
JU334182.1 - 1301 86
JU334183.1 - 1301 86
JU334184.1 - 1301 86
JU334185.1 - 1301 86
JU334186.1 - 1301 86
JU334187.1 - 1301 86
JU475312.1 - 1375 86
JU475313.1 - 1375 86
JU475314.1 - 1375 86
JU475315.1 - 1375 86
JV046861.1 - 1339 86
JV046862.1 - 1339 86
JV649437.1 - 1249 86

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Recommended Assay Macaca mulatta

Panther Classification:

Molecular Function -

general transcription factor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7dtsd:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0