This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh01092738_m1
See other FGF1 GE Assays ›
Gene Symbol:
FGF1
Gene Name:
fibroblast growth factor 1
Gene Aliases:
EGK_16969
Chromosome Location:
Chr.6: 139998490 - 140109214 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh01092738_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer:
435.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: FGF1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001266029.1 NP_001252958.1 304 104
XM_028849197.1 XP_028705030.1 399 104
XM_028849198.1 XP_028705031.1 436 104
XM_028849199.1 XP_028705032.1 1293 104
XM_028849200.1 XP_028705033.1 393 104
XM_028849201.1 XP_028705034.1 433 104
XM_028849202.1 XP_028705035.1 549 104
XM_028849203.1 XP_028705036.1 564 104
XM_028849204.1 XP_028705037.1 784 104
XM_028849205.1 XP_028705038.1 1782 104
XM_028849206.1 XP_028705039.1 563 104
GenBank mRNA JU333433.1 - 306 104
JU333434.1 - 304 104

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta

Panther Classification:

Molecular Function -

growth factor intercellular signal molecule

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-crt6c:80/100.66.78.36:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline