Assay Details
| Gene Symbol: TRIB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015111741.2 | XP_014967227.1 | 1696 | 93 | ||
| XM_015111742.2 | XP_014967228.1 | 634 | 93 | |||
| GenBank mRNA | JU320696.1 | - | 769 | 93 | ||
| JU475267.1 | - | 1664 | 93 | |||
| JV044779.1 | - | 1683 | 93 | |||
| JV636757.1 | - | 1684 | 93 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015111742.2 transcripts.
Set Membership: |
|

TRIB2
Genomic Map