This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh01125794_m1
See other FTSJ1 GE Assays ›
Gene Symbol:
FTSJ1
Gene Name:
FtsJ RNA 2'-O-methyltransferase 1
Gene Aliases:
EGK_20459
Chromosome Location:
Chr.X: 48465114 - 48480257 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh01125794_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: FTSJ1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001261002.2 NP_001247931.1 399 90
XM_015127313.2 XP_014982799.1 1267 90
XM_028841806.1 XP_028697639.1 1267 90
GenBank mRNA JU333759.1 - 555 90
JU333760.1 - 555 90
JU475008.1 - 571 90
JU475009.1 - 571 90
JV046631.1 - 399 90
JV577032.1 - 577 90

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Recommended Assay Macaca mulatta

Panther Classification:

Molecular Function -

RNA metabolism protein RNA methyltransferase nucleic acid metabolism protein

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8cb8z:80/100.66.72.77:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0