This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh01547530_m1
See other RBM41 GE Assays ›
Gene Symbol:
RBM41
Gene Name:
RNA binding motif protein 41
Gene Aliases:
EGK_20797
Chromosome Location:
Chr.X: 103830122 - 103887512 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh01547530_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RBM41
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001260581.1 NP_001247510.1 1111 80
XM_015127926.2 XP_014983412.2 2494 80
XM_015127927.2 XP_014983413.1 2487 80
XM_015127928.2 XP_014983414.1 2419 80
XM_028841742.1 XP_028697575.1 2490 80
XM_028841743.1 XP_028697576.1 2421 80
XM_028841744.1 XP_028697577.1 2119 80
GenBank mRNA JU335300.1 - 1110 80
JU476258.1 - 1111 80

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 3' Most Recommended Assay Macaca mulatta

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0