This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh01556193_m1
See other BRCA1 GE Assays ›
Gene Symbol:
BRCA1
Gene Name:
BRCA1 DNA repair associated
Gene Aliases:
-
Chromosome Location:
Chr.16: 54186473 - 54273232 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh01556193_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: BRCA1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001114949.1 NP_001108421.1 5466 59
XM_015119744.2 XP_014975230.1 6379 59
XM_015119745.2 XP_014975231.1 6373 59
XM_015119746.2 XP_014975232.1 6378 59
XM_015119747.2 XP_014975233.1 6378 59
XM_015119748.2 XP_014975234.1 6301 59
XM_028835632.1 XP_028691465.1 3061 59
XM_028835633.1 XP_028691466.1 2908 59
XM_028835634.1 XP_028691467.1 3085 59
XM_028835635.1 XP_028691468.1 2255 59
GenBank mRNA JU476929.1 - 5609 59

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Macaca mulatta

Panther Classification:

Molecular Function -

ubiquitin-protein ligase protein modifying enzyme

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0