This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh01566653_m1
See other DUSP15 GE Assays ›
Gene Symbol:
DUSP15
Gene Name:
dual specificity phosphatase 15
Gene Aliases:
-
Chromosome Location:
Chr.10: 67102651 - 67113961 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh01566653_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DUSP15
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015149276.2 XP_015004762.2 691 67
XM_015149278.2 XP_015004764.1 466 67
XM_015149279.2 XP_015004765.1 337 67
GenBank mRNA JU332919.1 - 406 67
JV046196.1 - 369 67
JV642287.1 - 407 67

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_015149278.2, XM_015149279.2 transcripts.

Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Macaca mulatta

Panther Classification:

Molecular Function -

protein phosphatase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline