This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh02788758_m1
See other UNC50 GE Assays ›
Gene Symbol:
UNC50
Gene Name:
unc-50 inner nuclear membrane RNA binding protein
Gene Aliases:
EGK_05597, GMH1
Chromosome Location:
Chr.13: 2378226 - 2393948 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh02788758_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: UNC50
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001195448.1 NP_001182377.1 385 78
XM_015112900.2 XP_014968386.2 467 78
GenBank mRNA AY680454.1 - 69 78
FS723120.1 - 401 78
JU333890.1 - 327 78
JV046694.1 - 308 78
JV046695.1 - 376 78

Back To Top

More Information


Additional Information:

This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.

Set Membership:

Amplicon length between 71 and 85 Recommended Assay Macaca mulatta

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0