This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh02788864_m1
See other ADGRG1 GE Assays ›
Gene Symbol:
ADGRG1
Gene Name:
adhesion G protein-coupled receptor G1
Gene Aliases:
EGK_12829, GPR56
Chromosome Location:
Chr.20: 44855566 - 44904737 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh02788864_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ADGRG1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001033020.1 NP_001028192.1 1968 71
XM_015126156.2 XP_014981642.1 2134 71
XM_015126157.2 XP_014981643.1 2280 71
XM_015126158.2 XP_014981644.1 2092 71
XM_015126161.2 XP_014981647.1 2092 71
XM_015126162.2 XP_014981648.1 2110 71
XM_015126163.2 XP_014981649.1 2162 71
XM_015126164.2 XP_014981650.1 2131 71
XM_015126165.2 XP_014981651.1 2161 71
XM_015126166.2 XP_014981652.1 2330 71
XM_015126167.2 XP_014981653.1 2077 71
XM_015126168.2 XP_014981654.1 2122 71
XM_015126169.2 XP_014981655.1 2143 71
XM_015126170.2 XP_014981656.1 2094 71
XM_015126171.2 XP_014981657.1 2561 71
XM_015126172.2 XP_014981658.1 2546 71
XM_028840341.1 XP_028696174.1 2025 71
XM_028840342.1 XP_028696175.1 2022 71
XM_028840344.1 XP_028696177.1 2227 71
XM_028840345.1 XP_028696178.1 2009 71
XM_028840346.1 XP_028696179.1 2034 71
XM_028840347.1 XP_028696180.1 2066 71
GenBank mRNA AY845352.1 - 1968 71
JU323293.1 - 2202 71
JU337110.1 - 2220 71
JU470897.1 - 2212 71
JU470898.1 - 2197 71
JU477855.1 - 2215 71
JV601061.1 - 2202 71
JV635411.1 - 2176 71

Back To Top

More Information


Additional Information:

This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.

Set Membership:

Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Recommended Assay Macaca mulatta

Panther Classification:

Molecular Function -

G-protein coupled receptor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2d9fq:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0