This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh02791973_m1
See other UBE2K GE Assays ›
Gene Symbol:
UBE2K
Gene Name:
ubiquitin conjugating enzyme E2 K
Gene Aliases:
EGK_15661, HIP2
Chromosome Location:
Chr.5: 38821227 - 38907212 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh02791973_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: UBE2K
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001260841.2 NP_001247770.1 349 116
XM_015138162.2 XP_014993648.1 582 116
XM_028848016.1 XP_028703849.1 485 116
GenBank mRNA JU327991.1 - 265 116
JU327992.1 - 265 116
JU472075.1 - 381 116
JV046905.1 - 350 116
JV668048.1 - 498 116

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_028848016.1 transcripts.
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0