Assay Details
| Gene Symbol: GLA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001093625.4 | XP_001093625.1 | 421 | 72 | ||
| GenBank mRNA | JU324188.1 | - | 304 | 72 | ||
| JU484534.1 | - | 258 | 72 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

GLA
Genomic Map