This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh02795256_m1
See other MGST3 GE Assays ›
Gene Symbol:
MGST3
Gene Name:
microsomal glutathione S-transferase 3
Gene Aliases:
-
Chromosome Location:
Chr.1: 23113693 - 23139536 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh02795256_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MGST3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001266781.2 NP_001253710.1 192 60
XM_015124516.2 XP_014980002.1 204 60
XM_015124521.2 XP_014980007.1 550 60
GenBank mRNA JU334562.1 - 119 60
JU475651.1 - 200 60
JV044624.1 - 196 60
JV601104.1 - 201 60
JV669196.1 - 201 60
KC734516.1 - 117 60

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Recommended Assay Macaca mulatta

Panther Classification:

Molecular Function -

transferase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline