Assay Details
| Gene Symbol: PLAC8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001258163.1 | NP_001245092.1 | 195 | 77 | ||
| XM_015138310.2 | XP_014993796.1 | 266 | 77 | |||
| GenBank mRNA | JU334857.1 | - | 178 | 77 | ||
| JU475880.1 | - | 195 | 77 | |||

PLAC8
Genomic Map