This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh02796436_m1
See other UCHL3 GE Assays ›
Gene Symbol:
UCHL3
Gene Name:
ubiquitin C-terminal hydrolase L3
Gene Aliases:
-
Chromosome Location:
Chr.17: 57327541 - 57420264 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh02796436_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: UCHL3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001266166.1 NP_001253095.1 459 65
XM_015121192.2 XP_014976678.1 710 65
XM_028837101.1 XP_028692934.1 710 65
XM_028837102.1 XP_028692935.1 710 65
XM_028837103.1 XP_028692936.1 710 65
XM_028837104.1 XP_028692937.1 710 65
XM_028837105.1 XP_028692938.1 710 65
XM_028837106.1 XP_028692939.1 710 65
GenBank mRNA JU321763.1 - 450 65
JV045679.1 - 459 65

Back To Top

More Information


Additional Information:

This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.

Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Macaca mulatta

Panther Classification:

Molecular Function -

cysteine protease

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7gdsv:80/100.66.79.88:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0