This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh02796732_m1
See other IAH1 GE Assays ›
Gene Symbol:
IAH1
Gene Name:
isoamyl acetate hydrolyzing esterase 1 (putative)
Gene Aliases:
-
Chromosome Location:
Chr.13: 99280747 - 99295100 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh02796732_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: IAH1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001193736.1 NP_001180665.1 599 105
XM_015111637.2 XP_014967123.1 942 105
XM_028831427.1 XP_028687260.1 510 105
XM_028831428.1 XP_028687261.1 367 105
GenBank mRNA JU334096.1 - 596 105
JU473721.1 - 580 105
JV045419.1 - 586 105
JV681241.1 - 590 105

Back To Top

More Information


Additional Information:

This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta

Panther Classification:

Molecular Function -

esterase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0