This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh02797497_m1
See other NAXD GE Assays ›
Gene Symbol:
NAXD
Gene Name:
NAD(P)HX dehydratase
Gene Aliases:
-
Chromosome Location:
Chr.17: 91997481 - 92069201 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh02797497_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NAXD
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015121446.2 XP_014976932.2 692 53
XM_015121447.2 XP_014976933.1 700 53
XM_015121448.2 XP_014976934.2 629 53
XM_015121449.2 XP_014976935.1 637 53
XM_028837311.1 XP_028693144.1 701 53
XM_028837312.1 XP_028693145.1 818 53
XM_028837313.1 XP_028693146.1 638 53
XM_028837314.1 XP_028693147.1 700 53
XM_028837316.1 XP_028693149.1 674 53
XM_028837317.1 XP_028693150.1 1123 53
XM_028837318.1 XP_028693151.1 795 53
XM_028837319.1 XP_028693152.1 586 53
XM_028837320.1 XP_028693153.1 708 53
GenBank mRNA JU329531.1 - 448 53
JV065541.1 - 551 53
JV655801.1 - 552 53

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 3' Most Recommended Assay Macaca mulatta

Panther Classification:

Molecular Function -

dehydratase lyase metabolite interconversion enzyme

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0