This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh02799768_m1
See other SLCO1A2 GE Assays ›
Gene Symbol:
SLCO1A2
Gene Name:
solute carrier organic anion transporter family member 1A2
Gene Aliases:
-
Chromosome Location:
Chr.11: 22647018 - 22815939 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh02799768_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SLCO1A2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001261711.1 NP_001248640.1 393 104
XM_015151116.2 XP_015006602.1 505 104
XM_015151117.2 XP_015006603.1 495 104
XM_015151118.2 XP_015006604.1 826 104
XM_015151119.2 XP_015006605.1 617 104
XM_015151120.2 XP_015006606.1 623 104
XM_015151122.2 XP_015006608.1 629 104
XM_015151123.2 XP_015006609.1 409 104
XM_015151125.2 XP_015006611.1 482 104
XM_028828875.1 XP_028684708.1 473 104
XM_028828876.1 XP_028684709.1 954 104
XM_028828877.1 XP_028684710.1 827 104
XM_028828878.1 XP_028684711.1 821 104
XM_028828879.1 XP_028684712.1 365 104
XM_028828880.1 XP_028684713.1 680 104
GenBank mRNA JU336011.1 - 393 104

Back To Top

More Information


Additional Information:

This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta

Panther Classification:

Molecular Function -

transporter

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0