This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh02801129_m1
See other DPY30 GE Assays ›
Gene Symbol:
DPY30
Gene Name:
dpy-30 histone methyltransferase complex regulatory subunit
Gene Aliases:
EGK_05206
Chromosome Location:
Chr.13: 76859179 - 76885538 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh02801129_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DPY30
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001261093.1 NP_001248022.1 131 118
XM_015112025.2 XP_014967511.1 244 118
XM_015112026.2 XP_014967512.1 244 118
XM_015112027.2 XP_014967513.1 251 118
XM_015112028.2 XP_014967514.1 247 118
XM_015112029.2 XP_014967515.1 298 118
XM_015112030.2 XP_014967516.1 360 118
XM_028831479.1 XP_028687312.1 244 118
XM_028831481.1 XP_028687314.1 263 118
XM_028831482.1 XP_028687315.1 244 118
XM_028831484.1 XP_028687317.1 243 118
GenBank mRNA JU320223.1 - 290 118
JU473496.1 - 128 118
JV048468.1 - 131 118
JV639282.1 - 302 118

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline