This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh02808310_g1
See other ATP5MG GE Assays ›
Gene Symbol:
ATP5MG
Gene Name:
ATP synthase membrane subunit g
Gene Aliases:
ATP5L
Chromosome Location:
Chr.14: 111457576 - 111465644 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh02808310_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ATP5MG
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001258127.1 NP_001245056.1 202 141
GenBank mRNA JU472390.1 - 202 141

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001258127.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Macaca mulatta

Panther Classification:

Molecular Function -

ATP synthase primary active transporter transporter

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0