This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh02818549_m1
See other GPR85 GE Assays ›
Gene Symbol:
GPR85
Gene Name:
G protein-coupled receptor 85
Gene Aliases:
EGK_14051
Chromosome Location:
Chr.3: 139356889 - 139366305 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh02818549_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GPR85
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001266820.1 NP_001253749.1 187 80
XM_015134730.2 XP_014990216.1 454 80
XM_015134731.2 XP_014990217.1 268 80
XM_015134732.2 XP_014990218.1 206 80
GenBank mRNA JU320024.1 - 187 80

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the NM_001266820.1, XM_015134730.2, XM_015134731.2, XM_015134732.2 transcripts.

Set Membership:

Amplicon length between 71 and 85 G-protein coupled receptor 3' Most Macaca mulatta

Panther Classification:

Molecular Function -

G-protein coupled receptor transmembrane signal receptor

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7dtsd:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0