This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh02818933_m1
See other ISLR2 GE Assays ›
Gene Symbol:
ISLR2
Gene Name:
immunoglobulin superfamily containing leucine rich repeat 2
Gene Aliases:
-
Chromosome Location:
Chr.7: 51134982 - 51144075 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh02818933_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ISLR2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015142689.2 XP_014998175.1 515 148
XM_015142691.2 XP_014998177.1 399 148
XM_028851123.1 XP_028706956.1 759 148
XM_028851124.1 XP_028706957.1 393 148
XM_028851125.1 XP_028706958.1 1276 148
XM_028851126.1 XP_028706959.1 387 148

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_015142689.2, XM_015142691.2, XM_028851123.1, XM_028851124.1, XM_028851125.1, XM_028851126.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Macaca mulatta

Panther Classification:

Molecular Function -

immunoglobulin superfamily cell adhesion molecule

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-4d4pq:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0