Assay Details
Gene Symbol: GREM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266108.1 | NP_001253037.1 | 221 | 96 | ||
GenBank mRNA | JV567203.1 | - | 215 | 96 | ||
JV625709.1 | - | 253 | 96 | |||
JV667455.1 | - | 221 | 96 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001266108.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
