This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh02826039_m1
See other MAMSTR GE Assays ›
Gene Symbol:
MAMSTR
Gene Name:
MEF2 activating motif and SAP domain containing transcriptional regulator
Gene Aliases:
-
Chromosome Location:
Chr.19: 48704694 - 48712480 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh02826039_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MAMSTR
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015124479.2 XP_014979965.1 2204 119
XM_015124480.2 XP_014979966.1 1581 119
XM_015124481.2 XP_014979967.1 2144 119
XM_015124482.2 XP_014979968.1 1521 119
XM_015124483.2 XP_014979969.1 2051 119
XM_015124485.2 XP_014979971.1 1658 119
XM_015124486.2 XP_014979972.1 1035 119
XM_015124487.2 XP_014979973.1 1239 119
GenBank mRNA JV648883.1 - 519 119

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0