This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh02826301_gH
See other ALDOA GE Assays ›
Gene Symbol:
ALDOA
Gene Name:
aldolase, fructose-bisphosphate A
Gene Aliases:
-
Chromosome Location:
Chr.20: 27285870 - 27292773 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh02826301_gH
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ALDOA
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_015125902.2 XP_014981388.1 1181 151
XM_015125903.1 XP_014981389.1 1316 151
XM_015125904.2 XP_014981390.1 1800 151
XM_028840863.1 XP_028696696.1 1189 151
XM_028840864.1 XP_028696697.1 1355 151
XM_028840865.1 XP_028696698.1 1443 151
GenBank mRNA JU470587.1 - 1287 151
JU470588.1 - 1432 151
JV043913.1 - 1131 151
JV043914.1 - 1172 151

Back To Top

More Information


Additional Information:

This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.

Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta

Panther Classification:

Molecular Function -

aldolase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0