This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Rh02828381_m1
See other GALE GE Assays ›
Gene Symbol:
GALE
Gene Name:
UDP-galactose-4-epimerase
Gene Aliases:
EGK_00373
Chromosome Location:
Chr.1: 200965506 - 200971095 on Build Mmul_10
Species:
Rhesus monkey
Species Specific ID (Flybase ID):
-
Assay ID Rh02828381_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GALE
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001261703.1 NP_001248632.1 362 112
XM_015130652.2 XP_014986138.1 566 112
XM_015130663.2 XP_014986149.1 568 112
XM_015130701.2 XP_014986187.1 566 112
XM_015130712.2 XP_014986198.1 564 112
XM_015130720.2 XP_014986206.1 565 112
XM_015130729.2 XP_014986215.1 584 112
XM_028844077.1 XP_028699910.1 1121 112
XM_028844092.1 XP_028699925.1 535 112
XM_028844095.1 XP_028699928.1 977 112
XM_028844100.1 XP_028699933.1 437 112
GenBank mRNA JU322689.1 - 362 112

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Recommended Assay Macaca mulatta

Panther Classification:

Molecular Function -

epimerase/racemase

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0